Order Entry
Switzerland
ContactUsLinkComponent
Drawer insert tray set, type B
Drawer insert tray set, type B
Catalog # WALD81012711
Drawer insert tray set, type B
Catalog # WALD81012711

Specifications

  • Description:
    Drawer inserts set of 7 pieces, for 1200 mm width drawer
  • W×D×H:
    1100×489×48 mm
  • Pk:
    1

Specifications

About this item

Drawer trays for fully equipping drawers and for underbench units with a depth of 1200 mm.