Orders LinkContactUsLinkComponent
Colour standards
Colour standards
Supplier:   Hach
Colour standards
Supplier:  Hach
Certificates
Colour standards
Colour standards
3 Options Available Below
Product Details & Documents
For accuracy checks in colour determinations.
  • Simple Accuracy Checks
  • Minimize waste with Voluette ampoules
  • Reliable results
The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.
Product Family Options
Product Information
  • Pack type
    Pk Info
    Pk
    Availability
    Price
  • 707-0009
    Standard colour solution, 15 Pt-Co units
    Standard colour solution, 15 Pt-Co units
    Bottle
    1× 1 litre
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 296.00
    SFr. 296.00
    1 * 1 L
       SFr. 296.00
  • 707-0001
    Standard colour solution, 500 Pt-Co units
    Standard colour solution, 500 Pt-Co units
    Voluette ampoule
    16× 10 ml
    160 mL
    Log in for availability
    Log in for availability
    1 * 160 mL
    1 * 160 mL - SFr. 153.00
    SFr. 153.00
    1 * 160 mL
       SFr. 153.00
  • 707-0002
    Standard colour solution, 500 Pt-Co units
    Standard colour solution, 500 Pt-Co units
    Bottle
    1× 1 litre
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 746.00
    SFr. 746.00
    1 * 1 L
       SFr. 746.00