Orders LinkContactUsLinkComponent
Bottles, square
Bottles, square
Bottles, square
Certificates
Bottles, square
2 Options Available Below
Product Details & Documents
Square bottles with clear-sighted thread.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Thread
    Colour
    W×D×H
    Availability
    Price
  • RIXI9-2200-0100-40
    Bottles, square
    Bottles, square
    100 ml
    RD 40
    Clear
    46×46×71 mm
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - SFr. 922.00
    SFr. 922.00
    1 * 480 ST
       SFr. 922.00
  • RIXI9-2250-0500-65
    Bottles, square
    Bottles, square
    500 ml
    RD 65
    Clear
    80×80×108 mm
    Log in for availability
    Log in for availability
    1 * 1.944 ST
    1 * 1.944 ST - SFr. 14’790.00
    SFr. 14’790.00
    1 * 1.944 ST
       SFr. 14’790.00