ContactUsLinkComponent
Yeast extract, Veggie™
Yeast extract, Veggie™
Catalog # 71279-3
CAS Number:  
Yeast extract, Veggie™
Catalog # 71279-3
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Veggie™ yeast extract
Specifications
About this item
Veggie yeast extract is a certified animal-free media component.
  • Ensures proper growth and maintenance of bacterial cells
  • Ideal for applications requiring minimal use of animal-derived materials
Used as a direct replacement for traditional yeast extract in bacterial growth media.