n-Heptane ≥99.5%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 5139-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (CH3(CH2)5CH3) (GC, corrected for water):min. 99.5 %
- Acidity (meq/g):max. 0.0005
- Residue after Evaporation:max. 0.0005 %
- Water (KF):max. 0.01 %
- at 210 nm:max. 1.0
- at 220 nm:max. 0.1
- at 230 nm:max. 0.02
- at 240 nm:max. 0.01
- at 250 nm:max. 0.005
Specification Test Results
About this item
:
:100.20 g/mol
Boiling Pt:98 °C
Density:0.68
Flash Pt:–4 °C
Storage Temperature:Ambient
UN:1206
ADR:3,
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544