Orders LinkContactUsLinkComponent
Titrators, SIMPLEX
Titrators, SIMPLEX
Supplier:   GERBER FUNKE
Titrators, SIMPLEX
Supplier:  GERBER FUNKE
Certificates
Titrators, SIMPLEX
Titrators, SIMPLEX
2 Options Available Below
Product Details & Documents
These SIMPLEX titrators consist of a polyethylene bottle on a plastic base.
  • Burette with automatic zero point adjustment
  • Precision titration by button control
Used for milk and cream.
Delivery information: Supplied with one 1 ml and one 25 ml pipette, one 200 ml Erlenmeyer flask.
Product Family Options
Product Information
  • Availability
    Price
  • GERB4520
    For milk, 0 - 25° SH
    For milk, 0 - 25° SH
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 199.00
    SFr. 199.00
    1 * 1 ST
       SFr. 199.00
  • GERB4521
    For cream, 0 - 40° SH
    For cream, 0 - 40° SH
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 194.00
    SFr. 194.00
    1 * 1 ST
       SFr. 194.00