Gauze compresses, Askina®
: B. Braun
Gauze compresses with cut edges for all types of classic wound care, as an ointment or medication carrier.
- 100% cotton
- Bleached without chlorine
- Can be cut to size
: DIN 14079 - VM 17, Ph. Eur.
Frequently Bought Together