Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Replacement Cables Electrophoresis Systems
VWR®, Replacement Cables Electrophoresis Systems
Catalog # 700-0853
Supplier:  VWR Collection
VWR®, Replacement Cables Electrophoresis Systems
Catalog # 700-0853
Supplier:  VWR Collection

Specifications

  • Description:
    Replacement cables with 4 mm male connection
  • Length:
    1 m
  • Pk:
    1 PAIR

Specifications

About this item

Replacement cables suitable for all PerfectBlue™ electrophoresis systems and blotting systems, and are compatible with the majority of commercially available power supplies.