Order Entry
Switzerland
ContactUsLinkComponent
VWR® PerfectBlue, Semi-dry electroblotters
VWR® PerfectBlue, Semi-dry electroblotters
Supplier:  VWR Collection
Certificates
undefined
VWR® PerfectBlue, Semi-dry electroblotters
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 700-1219
  • 700-1220
  • Model:
  • PerfectBlue S (10×10 cm)
  • PerfectBlue M (20×20 cm)
  • Buffer volume:
  • ~50 ml
  • ~200 ml
  • Active transfer area:
  • 100 cm³
  • 400 cm³
  • Transfer time:
  • 30 - 120 min
  • 30 - 120 min
  • Unit W×D×H:
  • 185×185×60 mm
  • 295×295×60 mm

Specifications

Frequently Bought Together