Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH buffer solutions, pH 1,679

pH buffer solutions, pH 1,679

pH buffer solution, 1,679, 250 mL
Catalog # 662-8101
Supplier:   Mettler - Toledo
pH buffer solutions, pH 1,679
Catalog # 662-8101
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, pH 1,679

pH buffer solution, 1,679, 250 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH buffer solution
  • pH value:
    1,679

Specifications