Order Entry
Switzerland
ContactUsLinkComponent
Mouse Recombinant EBI3 Sub-unit (IL-27/IL-35) (from E. coli)
Mouse Recombinant EBI3 Sub-unit (IL-27/IL-35) (from E. coli)
Certificates
undefined
Mouse Recombinant EBI3 Sub-unit (IL-27/IL-35) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity Assay: No test available, ED₅₀ ≤NA, Bioactivity ≥NA
  • Gene ID:
    O35228
  • Reconstitution instructions:
    Sterile 20 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    IL-27 EBI3 subunit|IL-35/EBI3|IL-35 EBI3 subunit
  • Protein/peptide name:
    EBI3 Sub-unit (IL-27 / IL-35)
  • Purity:
    ≥90%
  • Molecular weight:
    23 kDa
  • Sequence:
    MALVALSQPR VQCHASRYPV AVDCSWTPLQ APNSTRSTSF IATYRLGVAT QQQSQPCLQR SPQASRCTIP DVHLFSTVPY MLNVTAVHPG GASSSLLAFV AERIIKPDPP EGVRLRTAGQ RLQVLWHPPA SWPFPDIFSL KYRLRYRRRG ASHFRQVGPI EATTFTLRNS KPHAKYCIQV SAQDLTDYGK PSDWSLPGQV ESAPHKP
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together