Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant IFN-A 2A (from E. coli)
Human Recombinant IFN-A 2A (from E. coli)
Certificates
undefined
Human Recombinant IFN-A 2A (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity Assay: Viral CPE assay using EMC virus on A549 cells, ED₅₀ ≤0,005 ng/ml, Bioactivity ≥2,0×10⁸ units/mg
  • Gene ID:
    P01563
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Type I Interferon|Alpha 2
  • Protein/peptide name:
    IFN-A 2A
  • Purity:
    ≥95%
  • Molecular weight:
    19,4 kDa
  • Sequence:
    MCDLPQTHSL GSRRTLMLLA QMRKISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together