Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine

(S)-(-)-Nicotine

Glass bottle for solids, 10 g
Catalog # ROTH8746.1
Toxic
Environment
Marketsource
Supplier:   Roth Carl
(S)-(-)-Nicotine
Catalog # ROTH8746.1
Supplier:  Roth Carl
Marketsource
CAS Number:  54-11-5
(S)-(-)-Nicotine

Glass bottle for solids, 10 g

CAS Number:  54-11-5
Price
SFr. 43.10
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  • Merck Index:12,06611
  • Boiling Pt:247 °C (1013 hPa)
  • Melting Pt:–79 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:200-193-3
  • UN:1654
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.