Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(S)-(-)-Nicotine 99+%

(S)-(-)-Nicotine 99+%

Toxic
Environment
(S)-(-)-Nicotine 99+%
Certificates
(S)-(-)-Nicotine 99+%

4 Options Available Below

Product Details & Documents
Synonyms:  (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
  • CAS Number:54-11-5
  • Formula:C₁₀H₁₄N₂
  • MDL Number:MFCD00006369
  • Molecular Weight:162.23 g/mol
  • Merck Index:12,06611
  • Boiling Pt:247 °C (760 mmHg)
  • Melting Pt:–80 °C
  • Density:1.015 g/cm³
  • Flash Pt:101 °C
  • Storage Temperature:Ambient
  • EINECS:200-193-3
  • UN:1654
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5255 to 1.5285 (20°C, 589 nm)
  • Water:
    ≤0.5 % (K.F.)
  • GC:
    ≥99.0 %
  • Specific optical rotation:
    -140° to -152° (20°C, 589 nm) (c=2,absolute ethanol) (on anhydrous basis)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 40.70
    SFr. 40.70
    1 * 5 g
       SFr. 40.70

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 63.40
    SFr. 63.40
    1 * 25 g
       SFr. 63.40

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 162.00
    SFr. 162.00
    1 * 100 g
       SFr. 162.00

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 764.00
    SFr. 764.00
    1 * 500 g
       SFr. 764.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.