Mouse Recombinant VEGF-120 (from E. coli)
Vascular endothelial growth factor A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several alternatively spliced isoforms, with one being VEGF-120. VEGF-120 is an angiogenic factor that is expressed throughout endochondral bone development and is important during skeletogenesis.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: HUVEC proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
- Gene ID:Q00731-3
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:glioma-derived endothelial cell mitogen|VEGF-A|VPF
- Protein/peptide name:VEGF-120
- Purity:≥95%
- Molecular weight:14,2/28,4 kDa
- Sequence:MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together