


Pyridine ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:13,08060
Boiling Pt:12.4 %(V)
Melting Pt:–42 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
UN:1282
ADR:3,II
:79.1 g/mol
EINECS:203-809-9
:GHS07
:GHS08
:GHS02
Specification Test Results
- Autoignition Temperature:899 °F
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Assay (HClO₄-titr.):99.0 - 101.0 %
- Water (Karl Fischer):Max. 0.1 %
- APHA:Max. 50
- Vapor Pressure:10 mmHg ( 13.2 °C)
- Refractive Index:n20/D 1.509 (lit.)
- Explosion Limit:12.4 %
Specification Test Results
Frequently Bought Together