Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant LIF (from E. coli)
Human Recombinant LIF (from E. coli)
Certificates
undefined
Human Recombinant LIF (from E. coli)
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,2 ng/ml, Bioactivity ≥5,0×10⁶ units/mg
  • Gene ID:
    P15018
  • Reconstitution instructions:
    Sterile 10 mM acetic acid at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Leukocyte Inhibitory Factor
  • Protein/peptide name:
    LIF
  • Purity:
    ≥95%
  • Molecular weight:
    19,8 kDa
  • Sequence:
    MSPLPITPVN ATCAIRHPCH NNLMNQIRSQ LAQLNGSANA LFILYYTAQG EPFPNNLDKL CGPNVTDFPP FHANGTEKAK LVELYRIVVY LGTSLGNITR DQKILNPSAL SLHSKLNATA DILRGLLSNV LCRLCSKYHV GHVDVTYGPD TSGKDVFQKK KLGCQLLGKY KQIIAVLAQA F
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together