Mouse Recombinant IL-31 (from E. coli)
Fournisseur: Shenandoah Biotechnology
Certificats
À propos de cet article
Interleukin 31 (IL-31) is an immunoregulatory cytokine that is expressed by activated type 2 T helper (Th2) cells. IL-31 signals through a heterodimer receptor consisting of the IL-31 Receptor A (IL-31RA) and the oncostatin M receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 promotes allergic reactions and inflammatory skin diseases.
- High quality
- Low endotoxin
- Affordable
Attention: For research use only.
Spécifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q6EAL8
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:IL-31
- Purity:≥95%
- Molecular weight:15,7 kDa
- Sequence:MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Spécifications
Frequently Bought Together