Supelco® Organic Reference Standard, Nicotinamide (Niacinamide)
Catalog # 47865-U
Anbieter: Merck
CAS-Nummer:
Technische Daten
- Pk:1
- Standards:Analytical standard
- CAS No.:98-92-0
- Description:Nicotinamide (Niacinamide)
- Storage conditions:2...30 °C
- Hazard class description:H319
- Shipping conditions:STD
- MDL No.:MFCD00006395
- M-Clarity quality level:MQ100
- Synonym(s):Nicotinamide; NAM; NIC; Niacinamide; Nicotinic acid amide; Pyridine-3-carboxylic acid amide; Vitamin B₃; Vitamin PP
- Melting point:128 - 131 °C (lit.)
- Molecular weight:122,12
- Flash point:150 °C
Technische Daten
Über diesen Artikel
- GHS signal: GHS07
Caution: P264 - P280 - P305 + P351 + P338 - P337 + P313