Mouse Recombinant Rank Ligand (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Receptor activator of nuclear factor kappa-B Ligand (RANK Ligand) is a cell-bound marker related to the tumor necrosis factor (TNF) family of proteins. RANK Ligand plays a critical role in bone metabolism and osteoclast differentiation. T cell expression of RANK Ligand promotes dendritic cell maturation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: Ability to induce bioluminescence in engineered RANKL bioassay cells, ED₅₀ ≤50 ng/ml, Bioactivity ≥2,0×10⁴ units/mg
- Gene ID:O35235
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:OPGL|TNFSF11|ODF|TRANCE
- Protein/peptide name:Rank ligand
- Purity:≥95%
- Molecular weight:19,5 kDa
- Sequence:MPAMMEGSWLD VAQRGKPEAQ PFAHLTINAA SIPSGSHKVT LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPT DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISIQV SNPSLLDPDQ DATYFGAFKV QDID
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate and 20 mM Sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together