
Nicotinic acid (Niacin) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:59-67-6
:
Formula:C₆H₅NO₂
MDL Number:MFCD00006391
:123.11 g/mol
Merck Index:13,06552
Boiling Pt:258 °C (1013 hPa)
Melting Pt:142 °C
Density:1.473 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:193 °C
EINECS:200-441-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:18 g/l (20 °C)
- Melting Point:236.6 °C
- Molar Mass:123.11 g/mol
- Bulk Density:330 kg/m3
- Flash Point:193 °C
- Density:1.47 g/cm3 (20 °C)
- pH:2.7 (18 g/l, H2O, 20 °C)
- Ignition Point:365 °C Dust
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.18714.1000_TDS_EN ProductPage_818714
Product specification
msds_818714
SDS
Frequently Bought Together