Order Entry
Switzerland
ContactUsLinkComponent
Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.
Irritant
Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.
Certificates
undefined
Irritant
Nicotinic acid (Niacin), meets analytical specification of Ph.Eur.
Certificates

Specification Test Results

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    234 to 240 °C
  • Loss on drying:
    ≤1.0 %
  • Sulfated ash:
    ≤0.1 %
  • Solubility:
    (in boiling water) Soluble
  • Related substances:
    Passes test
  • Titration with NaOH:
    99.5 to 100.5 % (On dried substance)
  • Chloride (Cl):
    ≤200 ppm

Specification Test Results

Related Information

Frequently Bought Together