Order Entry
Switzerland
ContactUsLinkComponent
 
Rat Recombinant IFN-G (from E. coli)
 
undefined
Rat Recombinant IFN-G (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P01581
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Type II interferon|T cell interferon|MAF|immune interferon
  • UniProtKB:
    P01581
  • Protein/peptide name:
    IFN-gamma
  • Purity:
    ≥95%
  • Molecular weight:
    15,6/31,2 kDa
  • Sequence:
    MQGTLIESLE SLKNYFNSSS MDAMEGKSLL LDIWRNWQKD GNTKILESQI ISFYLRLFEV LKDNQAISNN ISVIESHLIT NFFSNSKAKK DAFMSIAKFE VNNPQIQHKA VNELIRVIHQ LSPESSLRKR KRSRC
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 100 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

 

Frequently Bought Together