Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant IL-13 (from E. coli)
Zertifikate
undefined
Human Recombinant IL-13 (from E. coli)

Technische Daten

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Gene ID:
    P35225
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    NC30
  • Protein/peptide name:
    IL-13
  • Purity:
    ≥95%
  • Molecular weight:
    12,6 kDa
  • Sequence:
    MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGQFN
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium citrate, pH 3,0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Technische Daten

Frequently Bought Together