Human Recombinant MCP-3 (from E. coli)
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
The research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell-based assays and cell culture.
: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
Monocyte chemotactic protein 3 (MCP-3), also called CCL7, is produced by macrophages and tumor cell lines. MCP-3 signals through the G protein-coupled receptors CCR1, CCR2, and CCR3. MCP-3 chemoattracts monocytes and regulates macrophage function during inflammation and metastasis.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
The research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell-based assays and cell culture.
: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
: For research use only.
- Catalog No:
- SHBT100-131-10UG
- SHBT100-131-100UG
- SHBT100-131-500UG
- SHBT100-131-1MG
- Protein/peptide name:
- MCP-3/CCL7
- MCP-3/CCL7
- MCP-3/CCL7
- MCP-3/CCL7
- Protein synonyms:
- MARC|CCL7
- MARC|CCL7
- MARC|CCL7
- MARC|CCL7
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Human
- Human
- Human
- Human
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- Q7Z7Q8
- Q7Z7Q8
- Q7Z7Q8
- Q7Z7Q8
- Sequence:
- QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
- QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
- QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
- QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL
- Protease-free:
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- Endotoxin LAL (EU/µg) ≤0.1
- UniProtKB:
- Q7Z7Q8
- Q7Z7Q8
- Q7Z7Q8
- Q7Z7Q8
- Biological activity:
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Bioactivity: No biological activity data is available at this time
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- 0,1% Trifluoroacetic Acid (TFA)
- Molecular weight:
- 9 kDa
- 9 kDa
- 9 kDa
- 9 kDa
- Reconstitution instructions:
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Sterile water at 0,1 mg/ml
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
- −20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
Product Family Options
- Pack typePk
Specifications
Cat. No.SHBT100-131-10UGProtein/peptide nameMCP-3/CCL7Protein synonymsMARC|CCL7Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ7Z7Q8SequenceQPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKLProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ7Z7Q8Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTSpecifications
Cat. No.SHBT100-131-100UGProtein/peptide nameMCP-3/CCL7Protein synonymsMARC|CCL7Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ7Z7Q8SequenceQPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKLProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ7Z7Q8Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTSpecifications
Cat. No.SHBT100-131-500UGProtein/peptide nameMCP-3/CCL7Protein synonymsMARC|CCL7Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ7Z7Q8SequenceQPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKLProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ7Z7Q8Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRTSpecifications
Cat. No.SHBT100-131-1MGProtein/peptide nameMCP-3/CCL7Protein synonymsMARC|CCL7Protein/peptide typeRecombinantProtein functionCytokineSpeciesHumanSourceE. coliConjugationUnconjugatedGene IDQ7Z7Q8SequenceQPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKLProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤0.1UniProtKBQ7Z7Q8Biological activityBioactivity: No biological activity data is available at this timePurity≥95%GradeRUOFormulation0,1% Trifluoroacetic Acid (TFA)Molecular weight9 kDaReconstitution instructionsSterile water at 0,1 mg/mlStorage conditions−20 °CShipping temperatureRT
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



