Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
8-Bromoadenine
Catalog # MOLEM22156050
Supplier:  Molekula
CAS Number:  
Irritant
8-Bromoadenine
Catalog # MOLEM22156050
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :214.02 g/mol
EINECS:230-225-1
 :GHS07
CAS Number:6974-78-3
Formula:C₅H₄BrN₅
MDL Number:MFCD00082518