Order Entry
Switzerland
ContactUsLinkComponent
 
Human Recombinant Eotaxin-3 (from E. coli)
 
undefined
Human Recombinant Eotaxin-3 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q9Y258
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    MIP-4 alpha|TSC-1|CCL26
  • Protein/peptide name:
    Eotaxin-3
  • Purity:
    ≥95%
  • Molecular weight:
    8,4 kDa
  • Sequence:
    TRGSDISKTC CFQYSHKPLP WTWVRSYEFT SNSCSQRAVI FTTKRGKKVC THPRKKWVQK YISLLKTPKQ L
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

 

Frequently Bought Together