Mouse Recombinant IL-3 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells. IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells. IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Gene ID:P01586
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:MCGF|P-cell stimulating factor|HCGF|Multi-CSF
- Protein/peptide name:IL-3
- Purity:≥95%
- Molecular weight:15,2 kDa
- Sequence:MDTHRLTRTL NCSSIVKEII GKLPEPELKT DDEGPSLRNK SFRRVNLSKF VESQGEVDPE DRYVIKSNLQ KLNCCLPTSA NDSALPGVFI RDLDDFRKKL RFYMVHLNDL ETVLTSRPPQ PASGSVSPNR GTVEC
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Related Information
Literature reference - PubMed ID 27786273 Literature reference - PubMed ID 29247574
Additional Product Details
Frequently Bought Together