Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethidium bromide

Ethidium bromide

Toxic
Health
Supplier:   EUROCLONE
Ethidium bromide
Supplier:  EUROCLONE
Certificates
Ethidium bromide

3 Options Available Below

Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • EINECS:214-984-6
  • UN:2811
  • ADR:6.1,I
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g