Human Recombinant IL-20 (from E. coli)
Anbieter: Shenandoah Biotechnology
Zertifikate
Über diesen Artikel
Interleukin 20 (IL-20) is structurally related to interleukin 10 (IL-10) and is produced by keratinocytes and monocytes. IL-20 acts through the STAT3 signaling pathway to regulate the proliferation of keratinocytes during epidermal inflammation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Technische Daten
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q9NYY1
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:IL-20
- Purity:≥95%
- Molecular weight:17,7/35,3 kDa
- Sequence:MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Technische Daten
Frequently Bought Together