Auftragserfassung
Switzerland
ContactUsLinkComponent
Phenanthren
Irritant
Environment
Phenanthren
Catalog # 123112M
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Environment
Phenanthren
Catalog # 123112M
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Packung:
    Glasampulle
  • VE:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :178.23 g/mol
Merck Index:12,07354
Boiling Pt:340 °C (1013 hPa)
Melting Pt:97…99 °C
Density:1.19 g/cm³ (20 °C)
Flash Pt:153 °C
Storage Temperature:Raumtemperatur
EINECS:201-581-5
UN:3077
ADR:9,III
 :GHS07
 :GHS09
CAS Number:85-01-8
Formula:C₁₄H₁₀
MDL Number:MFCD00001168