Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid

L(+)-Ascorbic acid

Marketsource
Supplier:   Roth Carl
L(+)-Ascorbic acid
Supplier:  Roth Carl
Marketsource
Certificates
L(+)-Ascorbic acid
L(+)-Ascorbic acid

3 Options Available Below

Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 19.10
    SFr. 19.10
    1 * 100 g
       SFr. 19.10

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - SFr. 26.30
    SFr. 26.30
    1 * 250 g
       SFr. 26.30

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 45.50
    SFr. 45.50
    1 * 500 g
       SFr. 45.50

    Specifications:
    Marketsource
    Marketsource