Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant FGF-6 (from E. coli)
Human Recombinant FGF-6 (from E. coli)
Certificates
undefined
Human Recombinant FGF-6 (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P10767
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    HSTF-2|HST-2|Heparin secretory-transforming protein 2|HBGF-6|Heparin-binding growth factor 6
  • Protein/peptide name:
    FGF-6
  • Purity:
    ≥95%
  • Molecular weight:
    18,9 kDa
  • Sequence:
    mgTRANNTLL DSRGWGTLLS RSRAGLAGEI AGVNWESGYL VGIKRQRRLY CNVGIGFHLQ VLPDGRISGT HEENPYSLLE ISTVERGVVS LFGVRSALFV AMNSKGRLYA TPSFQEECKF RETLLPNNYN AYESDLYQGT YIALSKYGRV KRGSKVSPIM TVTHFLPRI
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate and 50 mM sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together