

Manganese(IV) oxide ≥89.0%, powder, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1313-13-9
:
Formula:MnO₂
MDL Number:MFCD00003463
:86.94 g/mol
Merck Index:14,05730
Melting Pt:535 °C
Density:5.03 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:215-202-6
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (manganometric):≥ 89.0 %
- Substances insoluble in hydrochloric acid:≤ 5.0 %
- Sulphate (SO₄):≤ 0.1 %
- Ba (Barium):≤ 1.0 %
- Ca (Calcium):≤ 1.0 %
- Fe (Iron):≤ 1.5 %
- Loss on drying (105 °C):≤ 0.3 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05957.9025_TDS_EN ProductPage_105957
Product specification
msds_105957
SDS
Frequently Bought Together