Rat Recombinant FGF-basic (from E. coli)
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases.
- High Quality
- Low Endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: For research use only.
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases.
- High Quality
- Low Endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
: For research use only.
- Catalog No:
- SHBT300-02-10UG
- SHBT300-02-50UG
- SHBT300-02-100UG
- SHBT300-02-500UG
- SHBT300-02-1MG
- Protein/peptide name:
- FGF-basic
- FGF-basic
- FGF-basic
- FGF-basic
- FGF-basic
- Protein synonyms:
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Prostatropin|HBGF-2|FGF2
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Protein function:
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Cytokine
- Species:
- Rat
- Rat
- Rat
- Rat
- Rat
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- P13109
- P13109
- P13109
- P13109
- P13109
- Sequence:
- MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Protease-free:
- N
- N
- N
- N
- N
- Animal-free:
- N
- N
- N
- N
- N
- Nuclease-free:
- N
- N
- N
- N
- N
- Carrier-free:
- Y
- Y
- Y
- Y
- Y
- Endotoxin-free:
- N
- N
- N
- N
- N
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Endotoxin LAL (EU/µg) ≤1
- Biological activity:
- Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Purity:
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- ≥95%
- Grade:
- RUO
- RUO
- RUO
- RUO
- RUO
- Formulation:
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- 10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- Molecular weight:
- 16,4 kDa
- 16,4 kDa
- 16,4 kDa
- 16,4 kDa
- 16,4 kDa
- Reconstitution instructions:
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Sterile water at 0.1 mg/ml
- Storage conditions:
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- –20 °C
- Shipping temperature:
- RT
- RT
- RT
- RT
- RT
- Tested applications:
- Bioassay
- Bioassay
- Bioassay
- Bioassay
- Bioassay
Product Family Options
- Pack typePk
Specifications
Cat. No.SHBT300-02-10UGProtein/peptide nameFGF-basicProtein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP13109SequenceMPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5Molecular weight16,4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-02-50UGProtein/peptide nameFGF-basicProtein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP13109SequenceMPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5Molecular weight16,4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-02-100UGProtein/peptide nameFGF-basicProtein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP13109SequenceMPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5Molecular weight16,4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-02-500UGProtein/peptide nameFGF-basicProtein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP13109SequenceMPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5Molecular weight16,4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassaySpecifications
Cat. No.SHBT300-02-1MGProtein/peptide nameFGF-basicProtein synonymsProstatropin|HBGF-2|FGF2Protein/peptide typeRecombinantProtein functionCytokineSpeciesRatSourceE. coliConjugationUnconjugatedGene IDP13109SequenceMPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKSProtease-freeNAnimal-freeNNuclease-freeNCarrier-freeYEndotoxin-freeNEndotoxin levelLowEndotoxin contentEndotoxin LAL (EU/µg) ≤1Biological activityBioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mgPurity≥95%GradeRUOFormulation10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5Molecular weight16,4 kDaReconstitution instructionsSterile water at 0.1 mg/mlStorage conditions–20 °CShipping temperatureRTTested applicationsBioassay
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



