Mouse Recombinant EGF (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: BALB/c 3T3 cell proliferation, ED₅₀ ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
- Gene ID:P01132
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Urogastrone|URG
- Protein/peptide name:EGF
- Purity:≥95%
- Molecular weight:6,2 kDa
- Sequence:MNSYPGCPSS YDGYCLNGGV CMHIESLDSY TCNCVIGYSG DRCQTRDLRW WELR
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together