
Molybdenum(IV) sulphide 98.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1317-33-5
:
Formula:MoS₂
MDL Number:MFCD00003470
:160.07 g/mol
Merck Index:12,06318
Melting Pt:2375 °C
Density:4,8 g/cm³ (20 °C)
EINECS:215-263-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Grey to black Powder
- Assay:≥98.0 % (MoS2)
Specification Test Results
Frequently Bought Together