Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Vanadium(V) oxide ≥99.2%

Health
Irritant
Environment
Vanadium(V) oxide ≥99.2%
Certificates
Vanadium(V) oxide ≥99.2%
Vanadium(V) oxide ≥99.2%

3 Options Available Below

Product Details & Documents

Purity: 99.2% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Physical Form: Powder Density: 3.357 Melting Point: 690° Boiling Point: 1750° dec. Solubility: Slightly soluble in water. Soluble in acids and alkalies Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457

As an oxidation catalyst, in refractories, as a ceramics Colourant, in the manufacture of vanadium salts, and as a dye

Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Merck Index:14,9921
  • Boiling Pt:1750 °C (dec.)
  • Melting Pt:690 °C
  • Density:3.357
  • Storage Temperature:Ambient
  • EINECS:215-239-8
  • UN:2862
  • ADR:6.1,III
Product Family Options

Product Information