Vanadium(V) oxide 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:13,09987
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:13,09987
- Boiling Pt:1750 °C (760 mmHg)
- Melting Pt:690 °C
- Density:3.35 g/cm³
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to orange-brown Powder
- Assay:≥98.0 %
- Particle size:≤500 µm
- Chromium (Cr):≤200 ppm
- Iron (Fe):≤1000 ppm
- Silicon (Si):≤200 ppm
- Sodium oxide (Na2O):≤300 ppm
- Potassium oxide (K2O):≤300 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



