Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Calibration reference materials, alloys

Calibration reference materials, alloys

Supplier:   PerkinElmer
Calibration reference materials, alloys
Supplier:  PerkinElmer
Certificates
Calibration reference materials, alloys

5 Options Available Below

Product Details & Documents
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • PERCN5320110
    Alumel, three 50,8 mm pieces (154 °C)
    Alumel, three 50,8 mm pieces (154 °C)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 275.00
    SFr. 275.00
    1 * 1 ST
       SFr. 275.00

  • PERCN5320112
    Iron, three 50,8 mm pieces (780 °C)
    Iron, three 50,8 mm pieces (780 °C)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 275.00
    SFr. 275.00
    1 * 1 ST
       SFr. 275.00

  • PERCN5320113
    Nickel, three 50,8 mm pieces (354 °C)
    Nickel, three 50,8 mm pieces (354 °C)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 275.00
    SFr. 275.00
    1 * 1 ST
       SFr. 275.00

  • PERCN5320114
    Perkalloy, three 50,8 mm pieces (596 °C)
    Perkalloy, three 50,8 mm pieces (596 °C)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 358.00
    SFr. 358.00
    1 * 1 ST
       SFr. 358.00

  • PERCN5320201
    Calibration reference materials, alumel, nickel, perkalloy, iron, and cobalt
    Calibration reference materials, alumel, nickel, perkalloy, iron, and cobalt
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 722.00
    SFr. 722.00
    1 * 1 ST
       SFr. 722.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.