Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Protein Standard
Catalog # P8119-5ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Protein Standard
Catalog # P8119-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    80 mg/ml (HSA and gamma-globulins)
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • Description:
    Protein standard
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ200

Specifications

About this item