Zinc sulphate heptahydrate, granular USP, FCC, Macron Fine Chemicals™
:7446-20-0
:
:ZnSO₄·7H₂O
:MFCD00149894
:12,10293
:100 °C
:1.97 g/cm³ (20 °C)
:Ambient
:231-793-3
:3077
:9,III
:287.54 g/mol
:GHS05
:GHS07
:GHS09
- Environmentally Preferable:
- Meets U.S.P Requirements:—
- Food GMP Manufactured Product:—
- GMP Manufactured Product:—
- Meets F.C.C. Requirements:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Food Chemical:—
- Bulk Pharmaceutical Chemical:—
- Assay ((as ZnSO₄)):55.6 - 61.0 %
- Assay (ZnSO₄·7H₂O):99.0 - 108.7 %
- Identification A:Passes Test
- Identification B:Passes Test
- Appearance:Passes Test
- Acidity:Passes Test
- FCC - Alkalies and Alkaline Earths:≤0.5 %
- USP - Alkalies and Alkaline Earths:≤0.9 %
- Arsenic (As):≤3 ppm
- FCC - Iron (Fe):≤0.003 %
- FCC - Lead (Pb)(FCC):≤4 mg/kg
- USP - Lead (Pb):≤0.002 %
- FCC - Cadmium (Cd):≤2 mg/kg
- FCC - Mercury (Hg):≤5 ppm
- FCC - Selenium (Se):≤0.003 %
- Solubility:Passes Test
Frequently Bought Together