Order Entry
Switzerland
ContactUsLinkComponent
Mercury(II) sulphate, extra pure, Supelco®
Toxic
Health
Environment
Mercury(II) sulphate, extra pure, Supelco®
Supplier:  Merck
Certificates
Toxic
Health
Environment
Mercury(II) sulphate, extra pure, Supelco®
Supplier:  Merck

Specification Test Results

  • Identity:
    passes test
  • Chloride (Cl):
    ≤ 0.003 %
  • Fe (Iron):
    ≤ 0.005 %
  • Mercury (I) (as Hg):
    ≤ 0.1 %
  • Substances not reducing by formic acid (residue on ignition):
    ≤ 0.1 %
  • Ignition Point:
    450 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Frequently Bought Together