Order Entry
Switzerland
ContactUsLinkComponent
Mercury(II) sulphate, Supelco®
Toxic
Health
Environment
Mercury(II) sulphate, Supelco®
Catalog # 31014-100G
Supplier:  Merck
CAS Number:  
undefined
Toxic
Health
Environment
Mercury(II) sulphate, Supelco®
Catalog # 31014-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :296.65 g/mol
Merck Index:13,05914
Melting Pt:850 °C
Density:6.47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-992-5
UN:1645
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
 :GHS06
 :GHS08
 :GHS09
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047