Mercury(II) sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7135.1000
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Identity (Hg2+):passes test
- Molarity (K2Cr2O7):0.0198 - 0.0202
Specification Test Results
About this item
:
:296.65 g/mol
Merck Index:13,05914
Melting Pt:850 °C
Density:6.47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-992-5
UN:1645
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047