Order Entry
Switzerland
ContactUsLinkComponent
Zinc chloride ≥97%, TECHNICAL
Irritant
Corrosive
Environment
Zinc chloride ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Environment
Zinc chloride ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together