Copper(II) oxide
250 g
Catalog # MSPP200009554
Marketsource
Supplier: MARKET SOURCE PART PROCESS
Copper(II) oxide
Catalog # MSPP200009554
Supplier: MARKET SOURCE PART PROCESS
Marketsource
CAS Number: 1317-38-0
Jump to:
Product Details & Documents
Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
- CAS Number:1317-38-0
- Formula:CuO
- MDL Number:MFCD00010979
- Molecular Weight:79.55 g/mol
- Merck Index:13,02674
- Boiling Pt:2000 °C (1013 hPa)
- Melting Pt:1326 °C
- Density:4.9 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-269-1
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



