

Copper(II) oxide, fine wire ∼ 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1317-38-0
:
Formula:CuO
MDL Number:MFCD00010979
:79.55 g/mol
Merck Index:13,02674
Boiling Pt:2000 °C (1013 hPa)
Melting Pt:1300 °C
Density:6.2 g/cm3 (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:215-269-1
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (oxygen demand as CuO):≥ 55.0 %
- Chloride (Cl):≤ 0.005 %
- Total carbon (C):≤ 0.002 %
- Total nitrogen (N):≤ 0.002 %
- Total sulfur (as SO₄):≤ 0.005 %
Specification Test Results
Frequently Bought Together