Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Copper(I) oxide 97%

Corrosive
Irritant
Environment
Copper(I) oxide 97%
Certificates
Copper(I) oxide 97%

3 Options Available Below

Product Details & Documents
Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:13,02694
  • Boiling Pt:1800 °C (1013 hPa)
  • Melting Pt:1232 °C
  • Density:5,7 to 6.2 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Red-brown Powder
  • Titration Complexometric:
    ≥96.0 %
  • Iron (Fe):
    ≤500 ppm
  • Nickel (Ni):
    ≤100 ppm
  • Silver (Ag):
    ≤50 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 34.00
    SFr. 34.00
    1 * 25 g
       SFr. 34.00

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 71.10
    SFr. 71.10
    1 * 500 g
       SFr. 71.10

  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 275.00
    SFr. 275.00
    1 * 2,5 kg
       SFr. 275.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.