
Copper(I) iodide for synthesis, Sigma-Aldrich®
Catalog # 8.18311.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:0.08 g/l (18 °C)
- Melting Point:588 °C
- Molar Mass:190.45 g/mol
- Bulk Density:1400 kg/m3
- Boiling Point:1290 °C (1013 hPa)
- Density:5.62 g/cm3 (20 °C)
Specification Test Results
About this item
:
:190.45 g/mol
Merck Index:13,02692
Boiling Pt:1290 °C (1013 hPa)
Melting Pt:602 °C
Density:5.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-674-6
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ200
:GHS09
CAS Number:7681-65-4
:1335-23-5
Formula:CuI
MDL Number:MFCD00010978